Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-c-FOS Rabbit Polyclonal Antibody

Fournisseur: Biosensis
R-1751-50
BSENR-1751-50EA 312 EUR InStock
BSENR-1751-50
Anti-c-FOS Rabbit Polyclonal Antibody
Anticorps
This antibody is a nuclear phosphoprotein which forms a tight but non-covalently linked complex with the JUN/AP-1 transcription factor. In the heterodimer, FOS and JUN/AP-1 basic regions each seems to interact with symmetrical DNA half sites. On TGF-beta activation, forms a multimeric SMAD3/SMAD4/JUN/FOS complex at the AP1/SMAD-binding site to regulate TGF-beta-mediated signaling.

  • Has a critical function in regulating the development of cells
  • Has a important role in signal transduction, cell proliferation and differentiation

FUNCTION: Nuclear phosphoprotein which forms a tight but non-covalently linked complex with the JUP-1 transcription factor. In the heterodimer, FOS and JUP-1 basic regions each seems to interact with symmetrical DNA half sites. On TGF-beta activation, forms a multimeric SMAD3/SMAD4/JUN/FOS complex at the AP1/SMAD-binding site to regulate TGF-beta-mediated signaling. Has a critical function in regulating the development of cells destined to form and maintain the skeleton. It is thought to have an important role in signal transduction, cell proliferation and differentiation. In growing cells, activates phospholipid synthesis, possibly by activating CDS1 and PI4K2A. This activity requires Tyr-dephosphorylation and association with the endoplasmic reticulum. SUBUNIT: Heterodimer. Interacts with DSIPI; this interaction inhibits the binding of active AP1 to its target DNA. Interacts with MAFB. SUBCELLULAR LOCATION: Nucleus. INDUCTION: C-fos expression increases upon a variety of stimuli, including growth factors, cytokines, neurotransmitters, polypeptide hormones, stress and cell injury. SIMILARITY: Belongs to the bZIP family. Fos subfamily. SIMILARITY: Contains 1 bZIP domain (Ref: uniprot.org).



Immunocytochemistry (ICC): 1:2,000-1:10,000.

Immunohistochemistry (IHC): 1:20,000-1:50,000. This antibody has been shown to work on 4% PFA fixed mouse brain sections. Note that non-specific staining has been observed on

Concentration range for immunohistochemistry (IHC) and immunocytochemistry (ICC) is 1 to 2 µg/ml and for western blotting (WB) is 1 to 2 µg/ml. This antibody works on 4% PFA fixed mouse brain sections and detects bands between 50 to 65 kDa, which only appear in stimulated cells.

This antibody can be reconstituted with 50 µl sterile-filtered, ultrapure water, to achieve an antibody concentration of 1 mg/ml.
Store lyophilised, unopened vial at 2 to 8 °C or lower. After reconstitution, prepare aliquots and store at ‒20 °C for a higher stability. Avoid freeze-thaw cycles.

Type: Primary
Antigen: c-FOS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Cow, Horse, Human, Mouse, Pig, Rat

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité