Anti-GFP Rabbit Polyclonal Antibody
Fournisseur: Biosensis
R-1838-100
BSENR-1838-100EA 302 EUR InStock
BSENR-1838-100
Anti-GFP Rabbit Polyclonal Antibody
Anticorps
The green fluorescent protein (GFP) is a 27 kDa protein isolated originally from the jellyfish Aequoria victoria. It has an endogenous fluorochrome activity with excitation maximum at 395nm and emission maximum at 509 nm, which is similar to that of fluorescein. GFP can be expressed in fluorescent form in essentially any prokaryotic or eukaryotic cell.
This GFP rabbit antibody was made against a recombinant GFP construct originating from an Aequoria species which was engineered to improve spectral properties and prevent oligomerization (1). This form of GFP, referred to as AcGFP, is 94% identical to the eGFP developed by Tsien and co-workers. The antibody can be used to verify the expression, size and stability of both AcGFP and eGFP fusion proteins in western blotting experiments and to amplify GFP signals in tissues of transgenic animals.
Applications: Immunocytochemistry (1:2,000 - 1:5,000) and Western Blot (1:1,000 - 1:5,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité