PolTech Ultra DES Spray
Fournisseur: BASAN
2600041
BSNC2600041EA 31.3 EUR InStock
BSNC2600041
PolTech Ultra DES Spray
Produits nettoyants Désinfectants
Ready-to-use disinfectant cleaner (bactericidal, yeasticidal and virucidal against enveloped viruses) that allows the cleaning and disinfection of all washable surfaces (tables, worktops, railings, switches, etc.) in a single operation.
Particularly suitable for disinfecting surfaces in medical settings and in sectors such as public buildings, the food and non-food industry or the professional kitchen. Product approved for food contact.
Certifications: Bactericidal: EN1276, EN13697, Bactericide in medical settings: EN13727, Yeasticide: EN1650, EN13697, Yeasticide in medical settings: EN13624, Virucidal against enveloped viruses, Adenovirus and Coronavirus: EN14476.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité