ExoBrite™ True EV Membrane Stains
Fournisseur: Biotium
ExoBrite™ True
30129.EA 0 EUR InStock
30129. 30129-T
ExoBrite™ True EV Membrane Stains
Systèmes de liaison et de marquage des protéines
Lipophilic membrane dyes designed specifically for EV research and the best choice for general pan-EV staining.
  • Superior alternative to common membrane dyes like PKH, DiO, DiI, DiD
  • Near-complete coverage of EVs in a sample
  • Broad compatibility with EVs isolated from different sources
  • Compatible with antibody-co-staining
ExoBrite™ True EV membrane stains demonstrate higher (near-complete) coverage of EVs over competitor membrane dyes in fluorescence nanoparticle tracking analysis (fNTA). The dyes also offer clear differentiation of EVs from non-specific particles during flow detection. ExoBrite™ True EV Membrane Stains labeled all EV types tested to date.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité