Anti-ZAP70 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ZAP70/528]
Fournisseur: Biotium
BNUM0528-50
BTIUBNUM0528-50EA 417 EUR InStock
BTIUBNUM0528-50
Anti-ZAP70 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ZAP70/528]
Antilichamen
ZAP70 is a 70 kDa protein tyrosine kinase found in T-cells and natural killer cells.Control of this protein translation is via the IgVH gene. ZAP70 protein is expressed in leukemic cells of approximately 25% of chronic lymphocytic leukemia (CLL) cases as well.Anti-ZAP70 expression is an excellent surrogate marker for the distinction between the Ig-mutated (anti-ZAP70 negative) and Ig-unmutated (anti-ZAP70 positive) CLL subtypes and can identify patient groups with divergent clinical courses. The anti-ZAP70 positive Ig-unmutated CLL cases have been shown to have a poorer prognosis.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité