Anti-S100A9 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: S100A9/1011]
Fournisseur: Biotium
BNUM1011-50
BTIUBNUM1011-50EA 417 EUR InStock
BTIUBNUM1011-50
Anti-S100A9 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: S100A9/1011]
Anticorps
Recognizes a 14 kDa protein, identified as S100A9 (also known as Calgranulin B or MRP-14); expressed by granulocytes, monocytes and by tissue macrophages.The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. Altered expression of this protein is associated with the disease cystic fibrosis. This MAb reacts with neutrophils,More Product Information
Collapse All
Taille Cdt Numéro de catalogue VWR Quantité
50 μL 50 µl BTIUBNUM1011-50
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité