Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD63 Rat Monoclonal Antibody (ExoBrite™ 490/515)

Fournisseur: Biotium

P022-490-125EA 0 EUR InStock
P022-490-125
Anti-CD63 Rat Monoclonal Antibody (ExoBrite™ 490/515)
Anticorps
ExoBrite™ CD63 (Mouse) Flow antibody is validated for detection of mouse CD63 EV marker in isolated EVs. The antibody conjugates offer exceptional signal-to-noise and include colour options for the Pacific Blue™, FITC, and PE channels.

  • Optimal detection of mouse CD63 EV marker by flow cytometry
  • Validated for purified or bead-bound EVs
  • Bright signal and low background

Extracellular vesicles (EVs), including exosomes, are lipid-bound vesicles that are released from cells. EVs display specific surface proteins and can carry nucleic acids and other cargo, allowing them to transfer biological information between cells in different parts of the body. Therefore EVs are increasingly studied for their potential use in drug delivery and medical diagnostic applications.

The most common proteins used as EV markers are CD9, CD63, and CD81, members of the tetraspanin family. Tetraspanins are plasma membrane proteins with many proposed functions, including activation and sorting of other membrane proteins. They are also thought to play a role in the targeting of proteins to multivesicular bodies (MVBs) and exosomes. These tetraspanins are broadly expressed on many cell types and can therefore be detected on many types of EVs, but their expression levels vary depending on the cell type of origin.

Validated antibody for optimal detection of mouse CD63 EV marker in purified or bead-bound EVs by flow cytometry.

Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone:
Conjugation: ExoBrite™ 490/515
Epitope:
Host: Rat
Isotype:
IgG2a, kappa
Reactivity: Mouse

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité