Electroporation system, High Throughput ECM™ 630
Fournisseur: BTX
45-0654
BTXI45-0654EA 14380 EUR InStock
BTXI45-0654
Electroporation system, High Throughput ECM™ 630
Systèmes d'électroporation Électroporateurs
The High Throughput ECM™ 630 system offers a multi-well electroporation technology for processing multiple samples in seconds. Using the HT multi-well plates instead of traditional cuvettes, the researcher can transition from a single cuvette to either 25-well or 96-well electroporation by using one simple plate handler. This increases yields and the number of experimental runs in a single day. Experiments take seconds to run, allowing for quick and efficient optimisation of the electrical and biological parameters. Once optimised, samples are rapidly processed increasing yields and saving valuable time and money.
  • Can be used in bacteria, yeast and insect cell transformations
  • Screening of cDNA and siRNA library
  • High throughput 25 to 96-well
System is comprised of three components: muli-well plates, an HT plate handler and the ECM 630 generator.
* Includes ECM™ 630 generator, 25-well plates (2 mm gap, 6X), plate seals, HT-100 plate handler and a plate adaptor.
** Includes ECM™ 630 generator, 96-well Plates (2 mm gap, 2X), plate seals, HT-100 plate handler and a plate adaptor.
*** Includes ECM™ 630 generator, 630B safety stand, cuvettes 1 mm, 2 mm, 4 mm package of 30 (10 each), cuvette rack 660, enhancer 3000 probe, enhancer interface box, oscilloscope, cables, 25- well plates (2 mm gap, 6X), HT-200 plate pandler and a plate adaptor.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité