Holder, set to fix Interface and Woulff bottle on lab stand. To fix Interface I-300 / I-300 Pro, VacuBox, LegacyBox and Woulff bottle / valve unit on a lab stand, Rotavapor R-21x or Vacuum Pump V-7xx. Incl. holder for Interface and Woulff bottle, 2 M 1 * 1 SET
Fournisseur: BUCHI
Holder, set to fix Interface and Woulff bottle on lab stand. To fix Interface I-300 / I-300 Pro, VacuBox, LegacyBox and Woulff bottle / valve unit on a lab stand, Rotavapor R-21x or Vacuum Pump V-7xx. Incl. holder for Interface and Woulff bottle, 2 M 1 * 1 SET
COMMANDE
Développer tout / Réduire tout
Holder, set to fix Interface and Woulff bottle on lab stand. To fix Interface I-300 / I-300 Pro, VacuBox, LegacyBox and Woulff bottle / valve unit on a lab stand, Rotavapor R-21x or Vacuum Pump V-7xx. Incl. holder for Interface and Woulff bottle, 2 M 1 * 1 SET BUCH11065362 1 * 1 SET
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou