Chromogenic endotoxin testing reagents, Pyrochrome®
Fournisseur: ASSOCXIATES OF CAPE COD
C1500-5
CAPEC1500-5EA 1590 EUR InStock
CAPEC1500-5
Chromogenic endotoxin testing reagents, Pyrochrome®
Tests microbiologiques Tests sur les endoxines
Pyrochrome® sets the standard for quality and redefines the way chromogenic reagents should perform. Pyrochrome is a versatile quantitative reagent that is used to perform kinetic or endpoint assays. It is a sensitive and flexible reagent that can be used for testing in compliance with the USP, EP and JP bacterial endotoxins test chapter. Pyrochrome is offered with either Pyrochrome Buffer or for endotoxin-specific testing, with Glucashield® Buffer. Pyrochrome can be used with the new Pyros Kinetix® Flex tube reader at a 1:1 and economical 4:1 ratio.
  • Can achieve a maximum sensitivity of 0,001 EU/ml
  • Highly cost effective chromogenic reagent
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité