LAL-test, Pyrotell®-T, turbidimetric formulation
Fournisseur: ASSOCXIATES OF CAPE COD INR. I
T0051-5
CAPET0051-5EA 1420 EUR InStock
CAPET0051-5
LAL-test, Pyrotell®-T, turbidimetric formulation
Tests microbiologiques Tests sur les endoxines
Pyrotell®-T lysate is used to quantify endotoxin in kinetic turbidimetric tests.
Pyrotell-T lysate can be used with the Pyros Kinetix® tube reader and also incubating microplate readers. When used with the pyros kinetix tube reader, Pyrotell-T gives a highly economic, flexible and sensitive LAL assay.
Pyrotell-T lysate can be used for a wide variety of endotoxin tests, ranging from standard water testing to samples requiring high sensitivity, such as intrathecal products and those requiring high dilutions to overcome interference.
Information de livraison: Pyrotell-T lysate is supplied in individual multi-test vials. Each vial contains sufficient reagent for approximately 96 tests.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité