Overhead stirrer, R100
Fournisseur: CAT (ZIPPERER GMBH)
60218-00
CATA60218-00EA 3460 EUR InStock
CATA60218-00
Overhead stirrer, R100
Agitateurs Agitateurs à tige
The powerful pilot plant stirrers for heavy duty jobs. The R100-series is designed for stirring applications up to 200 litres; ideal for medium and high viscous media −100000 mPa·s. Powerful microprocessor controlled (fuzzy logic) stirrers for optimum performance and accuracy, with a hollow shaft and precision chuck of 10,5 mm. The electronic overload and overheat protection shuts off the unit automatically avoiding damage to the stirrer.
  • Bright back-lit digital display shows all relevant system data (power input, power output and torque) plus simultaneous display of set and actual motor speed or time
  • Display also gives readout of the absolute or relative torque measurements as well as the power output in watts at the shaft
  • Programmable torque limits as well as maximum and minimum motor speeds
  • RS232-interface and analogue input for easy remote speed control and analogue output for recording as well as a programmable switchoff timer
R100CT: Stirrer has maximum torque of 600 Ncm, Speed range 10 to 500 min⁻¹ with soft start and soft stop logic. Ideal for medium and high viscous media (10000 to 100000 mPa·s)
R100CL: Same as R100CT but has speed range from 40 to1100 min⁻¹ and a higher torque of 250 Ncm; ideal for medium to high viscous media (10000 to 100000 mPa·s).
R100C: The universal laboratory stirring motor for rheometric tests. Same as R100CT but with a low speed range 30 to 2000 min⁻¹ and high torque of 150 Ncm. A multi-step safety system carries out a self-test of all safety functions and detects and protects against overload as well as over-temperature.
R100SD: Powerful, economical overhead stirrer with 4-digit speed display. The microprocessor controlled speed is steplessly adjustable from 40 to 2000 min⁻¹. The softstart and soft-stop logic prevents liquid splashing. The unit is equipped with an extra long life high performance brush motor. Max. torque 100 Ncm.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité