ELISA Kit, Anti-Ovalbumin IgG1 (Mouse)
Fournisseur: Cayman Chemical
500830-96
CAYM500830-96EA 746 EUR InStock
CAYM500830-96
ELISA Kit, Anti-Ovalbumin IgG1 (Mouse)
Tests Tests ELISA
For the measurement of anti-ovalbumin of the IgG1 isotype in mouse plasma and serum.
  • Measure mouse anti-OVA IgG1 in mouse plasma, serum, or BAL fluids
  • Employs an affinity-purified mouse anti-OVA IgG1 polyclonal antibody standard for accurate results
  • Assay Range: 1,56 to 200 ng/ml with an LLOQ of 1,56 ng/ml
  • Rapid assay, get results in under four hours
Cayman's ovalbumin IgG1 (mouse) ELISA kit is an immunometric (i.e., 'sandwich') assay which can be used to measure anti-ovalbumin of the IgG₁ isotype in mouse plasma and serum without prior sample purification.
Information de livraison: Contents: antibody, standard, buffer, polysorbate 20, substrate solution, stop solution, plate, and cover sheet
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité