COX Colorimetric Inhibitor Screening Assay Kit
Fournisseur: Cayman Chemical
701050-192
CAYM701050-192EA 1340 EUR InStock
CAYM701050-192
COX Colorimetric Inhibitor Screening Assay Kit
Tests Tests ELISA
A colorimetric assay for the screening of isozyme-specific inhibitors.
  • Screen for inhibitors of ovine COX-1 or human COX-2
  • Measure the peroxidase component of the COX
  • Assay 41 samples for COX-1 and 41 samples for COX-2 in duplicate
  • Plate-based colorimetric measurement (590 nm)
Cayman's COX Colorimetric Inhibitor Screening Assay measures the peroxidase component of cyclooxygenases. The peroxidase activity is assayed colorimetrically by monitoring the appearance of oxidized N,N,N',N'-tetramethyl-p-phenylenediamine (TMPD) at 590 nm. Inhibition of COX activity, measured by TMPD oxidation, by a variety of selective and nonselective inhibitors, shows changed potencies similar to those observed with other in vitro methods. The COX Colorimetric Inhibitor Screening assay includes both ovine COX-1 and human recombinant COX-2 enzymes in order to screen isozyme-specific inhibitors. Cayman's COX Colorimetric Assay is a time saving tool for screening vast numbers of inhibitors.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité