Catalase Assay Kit
Fournisseur: Cayman Chemical
707002-480
CAYM707002-480EA 973 EUR InStock
CAYM707002-480
Catalase Assay Kit
Tests Tests enzymatiques
For the measurement of catalase activity in plasma, serum, erythrocyte lysates, tissue homogenates, and cell lysates.
  • Assay 40 samples in duplicate
  • Measure catalase activity down to 2 U/ml
  • Plate-based colorimetric measurement (540 nm)
Catalase is an ubiquitous antioxidant enzyme that is present in most aerobic cells. It is involved in the detoxification of H2O2, a ROS that is a toxic product of both normal aerobic metabolism and pathogenic ROS production. Cayman’s Catalase Assay utilises the peroxidatic function of catalase for determination of enzyme activity.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité