Passage Reagent Group™ Reagents
Fournisseur: CELL SYSTEMS CORPORATION
4Z0-800
CELS4Z0-800EA 164.11 EUR InStock
CELS4Z0-800
Passage Reagent Group™ Reagents
Milieux de culture cellulaire
Optimize the plating, passaging and freezing of cell cultures with Cell Systems Reagents. Growth supplements and antibiotic products are also available to ensure each experiment is initiated in an optimal environment.
  • High quality specialised reagents
  • Always available
  • Overnight delivery
Passage Reagent Group™ is a matched set of Cell Systems Certified™ reagents for releasing cells from culture for subculture or freezing.
This PRG Group™ is qualified for use with all Cell Systems Certified Mediums. The PRG contains three parts: PRG-1™ (EDTA-dPBS Solution), PRG-2™ (Trypsin/EDTA-dPBS Solution) and PRG-3™(Trypsin Inhibitor-dPBS Solution) each in a 100 ml bottle. Cell Systems media and reagents are Sterile, made with WFI and all components are cGMP and ISO Compliant.
Cell membranes are materially and cumulatively damaged whenever cells are exposed to serine proteases, physically manipulated, centrifuged, and/or frozen. Use of the PRG greatly minimises damage and stress to cells during passage or freezing of cell cultures.
PRG-1™ (EDTA-dPBS Solution): The chelating agent EDTA in PRG-1™ prepares the cells for PRG-2™ processing.
PRG-2™: Contains highly purified trypsin, allowing a substantial reduction (40%) in the amount of enzyme required to detach cells compared to typical commercial trypsin solutions. The trypsin in PRG-2™ is inactivated by trypsin inhibitors in the PRG-3™ formulation, preventing nonspecific protease damage to cell membranes after detachment.
PRG-3™: Inactivates the protease in PRG-2™ and stabilises the cell membranes for passaging and freezing. PRG-3™ is engineered to work together with Attachment Factor™ (Cat. #4Z0-210) to stabilise the cell membrane and promote spreading and the establishment of correct polarity after subculture.
Our Passage Reagent System™ (cat. #4Z0-840), adds AttachmentFactor™ to the PRG Group™.
Certifications: Cell Systems media and reagents are Sterile, made with WFI and all components are cGMP and ISO Compliant.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité