Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Primary Human Glomerular Microvascular Endothelial Cells

Fournisseur: CELL SYSTEMS CORPORATION

ACBRI128
CELSACBRI128EA 861.32 EUR InStock
CELSACBRI128
Primary Human Glomerular Microvascular Endothelial Cells
Cellules
Our primary human cells offer unmatched consistency and a more biologically relevant tool than other cell types. Our cells are antibody-free and come from a deep inventory to help researchers with reproducibility and improved human insights.

  • Highly proliferative and healthy (~85% viability)
  • Antibody free and more biologically relevant
  • Offered with high cell count (~1 million cells per vial)
  • Well-cited in scientific journals
  • Ideal for 2D and 3D cultures, microfluidic devices and co-cultures

Primary Human Glomerular Microvascular Endothelial Cells (ACBRI128) were initiated by decapsulated decapsulated glomeruli isolated from normal human kidney cortical tissue. This cell is antibody free. Each vial will initiate a 25 to 75 cm² cell culture. These primary cells are isolated, frozen and qualified using Cell Systems Certified Media and Reagents--which are specified for use in the research environment. Cell Systems primary cells are exclusively from normal, site-specific human tissue specimen, and available in frozen vial or proliferating flask.

These cells were originated using Cell Systems Complete Serum-Free Medium (SF-4Z0-500), and subsequently grown and passaged in Cell Systems Complete Medium (4Z0-500). They are available at Passage 3 (<12 cumulative population doubling) cryopreserved in Cell Systems Cell Freezing Medium™ (4Z0-705). This vial will initiate a Passage 4 cell culture in a 75 cm² flask. Each vial contains more than 1×106 cells.

Each vial of cells is shipped with vials of Bac-Off® (antibiotic) and CultureBoost™ (animal derived growth factors) or CultureBoost-R™ (human recombinant growth factors) at no additional cost.

Standard Tests:
HIV Serologic Test (donor level HIV AB EIA) -Negative
HIV PCR Test (frozen cell pool by CLIA Licensed Clinical Lab) - Negative
Hepatitis B (HBV) PCR Test (frozen cells) - Negative
Hepatitis C (HCV) PCR Test (frozen cells) - Negative
Mycoplasma spp. (frozen cells) (ATCC method by CLIA Licensed Clinical Lab) - Negative.

Characterisation:
Inducible expression of CD 62E (E-Selectin) <2% positive by immunofluorescence
Cytoplasmic Vimentin Intermediate Filaments >98% positive by immunofluorescence
Cytoplasmic Alpha-Actinin Microfilaments >98% positive by immunofluorescence
Cytoplasmic Fibronectin >98% positive by immunofluorescence.

Information de livraison: Shipped on dry ice.

Attention: For research use only.


Commander maintenant

 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité