Stationary Tank System for Lubricants, MULTI-Tank
Fournisseur: CEMO GMBH
7536
CEMG7536EA 1020 EUR InStock
CEMG7536
Stationary Tank System for Lubricants, MULTI-Tank
Récipients Récipients rectangulaires
Tank from HDPE with general construction inspection approval with rigidly connected sheet steel pallet for handling with a forklift or pallet truck from any of the four sides.
  • Tank as described above with integral sheet steel pallet for driving under with a forklift or hand forklift from any of four sides
  • Approved for transport according to the ADR, but without a pump fitted
  • Lower centre of gravity increases stability and improves handling/ transport
  • For storage of diesel, biodiesel without a bund, even in water-protected areas
  • Certified for installation indoors
Individual tank with integral galvanised sheet steel bund, standard accessories such as filling contents gauge, optical leakage detector device, integral carrying handle, no pump.
Commander maintenant
COMMANDE
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité