Drum depot
Fournisseur: CEMO GMBH
8933
CEMG8933EA 2210 EUR InStock
CEMG8933
Drum depot
Stockage des fûts
Drum depots, made of PE, suitable for safe storage of 200 L drums indoors or outdoors. They are approved for the storage of water-polluting and aggressive liquids.
  • Hood with four cut-outs for ground anchoring
  • High chemical resistance to aggressive media
  • Locable and has two storage compartments on the inside side wall
  • Two smooth-running, rugged, yet flexible and PE blinds for access from both sides
These depot and has collection tray which can be driven under on both sides, perforated plate and hood in polyethylene (PE).
Commander maintenant
COMMANDE
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité