Alkaline detergent, RBS IND 470
Fournisseur: CHEMICAL-PRODUCTS
RBSIND470
CHEPRBSIND470EA 87.8 EUR InStock
CHEPRBSIND470
Alkaline detergent, RBS IND 470
Produits nettoyants Détergents et Nettoyants de Surfaces
Concentrated alkaline liquid detergent for radioactive decontamination of parts and surfaces made of glass and stainless steel.
  • Cleans and decontaminates
  • Halogen-free
  • high emulsifying and detergency power
RBS IND 470 is recommended for manual cleaning and soaking of parts, surfaces and containers made of stainless steel, glass and plastics. It is suitable for aqueous cleaning processes in laboratories and industry (nuclear power station, cyclotrons and others institutions that use to handle radioactive products).
Informations de commande: Available in box of 4 × 5 litres. Please contact Avantor for details.
Attention: Not recommended for use on white metals.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité