Laminar flow cabinets, CrossFlow CLF series
Fournisseur: CleanAir by Baker
1560401 1560402 1560403 1560404 1560405 1560406
CLEA1560401EA 8403 EUR InStock
CLEA1560401 CLEA1560402 CLEA1560403 CLEA1560404 CLEA1560405 CLEA1560406
Laminar flow cabinets, CrossFlow CLF series
Hottes et coffrets Hottes à flux laminaire
The CrossFlow series is a high quality crossflow cabinet range for handling non-hazardous materials that require a clean working environment. The ergonomically designed HEPA filtered horizontal laminar airflow (crossflow) creates an ISO Class 5 (14644-1) / GMP Class A (Annex 1) work area with low noise level and prevents ambient air from entering the work area, ensuring product protection.
  • Microprocessor flow control
  • User-friendly control panel for intuitively operating
  • HEPA filter H14 (efficiency of 99,995% MPPS) and pre-filter (ISO 16890)
  • Energy saving
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité