,,1 component: Metolachlor ESA sodium salt [CAS:947601-85-6] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store in a freezer at -18C or below,, 1 * 1 mL
Fournisseur: CUSTOM MADE CHEMICALS LAB
,,1 component: Metolachlor ESA sodium salt [CAS:947601-85-6] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store in a freezer at -18C or below,, 1 * 1 mL
New Product
COMMANDE
Développer tout / Réduire tout
,,1 component: Metolachlor ESA sodium salt [CAS:947601-85-6] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store in a freezer at -18°C or below,, 1 * 1 mL CMPLRD0025151 1 * 1 mL
New Product
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou