,,1 component: Chloridazon-desphenyl [CAS:6339-19-1] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store in a refrigerator at temperatures between 2C to 8C,, 1 * 1 mL
Fournisseur: CUSTOM MADE CHEMICALS LAB
,,1 component: Chloridazon-desphenyl [CAS:6339-19-1] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store in a refrigerator at temperatures between 2C to 8C,, 1 * 1 mL
New Product
COMMANDE
Développer tout / Réduire tout
,,1 component: Chloridazon-desphenyl [CAS:6339-19-1] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store in a refrigerator at temperatures between 2°C to 8°C,, 1 * 1 mL CMPLRD0379061 1 * 1 mL
New Product
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou