,,1 component: Chloridazon-methyl-desphenyl [CAS:17254-80-7] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store under normal laboratory conditions, at temperatures between 15C to 25C,, 1 * 1 mL
Fournisseur: CUSTOM MADE CHEMICALS LAB
,,1 component: Chloridazon-methyl-desphenyl [CAS:17254-80-7] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store under normal laboratory conditions, at temperatures between 15C to 25C,, 1 * 1 mL
New Product
COMMANDE
Développer tout / Réduire tout
,,1 component: Chloridazon-methyl-desphenyl [CAS:17254-80-7] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store under normal laboratory conditions, at temperatures between 15°C to 25°C,, 1 * 1 mL CMPLRD0782191 1 * 1 mL
New Product
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou