,, ,,1 component: Chlorothalonil metabolite R417888 sodium [CAS:N/A] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store under normal laboratory conditions, at temperatures between 15°C to 25°C,, 1 * 1 mL
Fournisseur: CUSTOM MADE CHEMICALS LAB
New Product
COMMANDE
Développer tout / Réduire tout
,, ,,1 component: Chlorothalonil metabolite R417888 sodium [CAS:N/A] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store under normal laboratory conditions, at temperatures between 15°C to 25°C,, 1 * 1 mL CMPLRD0923401 1 * 1 mL
New Product
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou