,,1 component: Iodobenzene as Iodides [CAS:591-50-4] 500ug/ml in Toluene ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 12 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15C to 25C,,,, 1 * 100 mL
Fournisseur: CUSTOM MADE CHEMICALS LAB
,,1 component: Iodobenzene as Iodides [CAS:591-50-4] 500ug/ml in Toluene ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 12 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15C to 25C,,,, 1 * 100 mL
New Product
COMMANDE
Développer tout / Réduire tout
,,1 component: Iodobenzene as Iodides [CAS:591-50-4] 500ug/ml in Toluene ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 12 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15°C to 25°C,,,, 1 * 100 mL CMPLRD109272L1 1 * 100 mL
New Product
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou