19 components: 1000mg/l each of Al B Ba Ca Cd Co Cr Cu Fe K Mg Mn Na Ni Pb S Ti V Zn in HNO3 5% ,, CRM, ISO 17034 and ISO 17025,, Conservation 24 months. (12 months after opening),, Ambient Temp.,, 1 * 100 mL
Fournisseur: CUSTOM MADE CHEMICALS LAB
19 components: 1000mg/l each of Al B Ba Ca Cd Co Cr Cu Fe K Mg Mn Na Ni Pb S Ti V Zn in HNO3 5% ,, CRM, ISO 17034 and ISO 17025,, Conservation 24 months. (12 months after opening),, Ambient Temp.,, 1 * 100 mL
COMMANDE
Développer tout / Réduire tout
19 components: 1000mg/l each of Al B Ba Ca Cd Co Cr Cu Fe K Mg Mn Na Ni Pb S Ti V Zn in HNO3 5% ,, CRM, ISO 17034 and ISO 17025,, Conservation 24 months. (12 months after opening),, Ambient Temp.,, 1 * 100 mL CMPLRM029054L1 1 * 100 mL
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou