5 components: Ammonium (NH4+) 100mg/l Potassium (K+) 100mg/l Sodium (Na+) 100mg/l Lithium (Li+) 20mg/l Ethanolamine (for Inorganics) (C2H7NO) 100mg/l in Water ,, CRM, ISO 17034 and ISO 17025,, Conservation 12 months. (6 months after opening), 1 * 500 mL
Fournisseur: CUSTOM MADE CHEMICALS LAB
5 components: Ammonium (NH4+) 100mg/l Potassium (K+) 100mg/l Sodium (Na+) 100mg/l Lithium (Li+) 20mg/l Ethanolamine (for Inorganics) (C2H7NO) 100mg/l in Water ,, CRM, ISO 17034 and ISO 17025,, Conservation 12 months. (6 months after opening), 1 * 500 mL
COMMANDE
Développer tout / Réduire tout
5 components: Ammonium (NH4+) 100mg/l Potassium (K+) 100mg/l Sodium (Na+) 100mg/l Lithium (Li+) 20mg/l Ethanolamine (for Inorganics) (C2H7NO) 100mg/l in Water ,, CRM, ISO 17034 and ISO 17025,, Conservation 12 months. (6 months after opening), 1 * 500 mL CMPLRM092682L5 1 * 500 mL
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou