5 components: 100mg/l each of As Ni Pb Sb Si in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 500 mL
Fournisseur: CUSTOM MADE CHEMICALS LAB
5 components: 100mg/l each of As Ni Pb Sb Si in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 500 mL
COMMANDE
Développer tout / Réduire tout
[EN]MIXTURE 5 COMPONENTS 1 * 500 mL CMPLRM093955L5 1 * 500 mL
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou