,,4 components: 500mg/l each of Chlorides (Cl-) Nitrates (NO3-) Phosphates (PO43-) Sulphates (SO42-) in Water ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months. (12 months after opening),, ,,Ambient Temp.,, 1 * 200 mL
Fournisseur: CUSTOM MADE CHEMICALS LAB
,,4 components: 500mg/l each of Chlorides (Cl-) Nitrates (NO3-) Phosphates (PO43-) Sulphates (SO42-) in Water ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months. (12 months after opening),, ,,Ambient Temp.,, 1 * 200 mL
New Product
COMMANDE
Développer tout / Réduire tout
,,4 components: 500mg/l each of Chlorides (Cl-) Nitrates (NO3-) Phosphates (PO43-) Sulphates (SO42-) in Water ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months. (12 months after opening),, ,,Ambient Temp.,, 1 * 200 mL CMPLRM096013L2 1 * 200 mL
New Product
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou