,,9 components: 1000mg/l each of Cr Co Ni As Cd Sb Pb Ti Zn in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months. (12 months after opening),, ,,Ambient Temp.,, 1 * 100 mL
Fournisseur: CUSTOM MADE CHEMICALS LAB
,,9 components: 1000mg/l each of Cr Co Ni As Cd Sb Pb Ti Zn in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months. (12 months after opening),, ,,Ambient Temp.,, 1 * 100 mL
COMMANDE
Développer tout / Réduire tout
,,9 components: 1000mg/l each of Cr Co Ni As Cd Sb Pb Ti Zn in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months. (12 months after opening),, ,,Ambient Temp.,, 1 * 100 mL CMPLRM099931L1 1 * 100 mL
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou