,,2 components: 1000mg/l each of Si Ti in HNO3 5% HF 0.2% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15°C to 25°C,,,, 1 * 100 mL
Fournisseur: CUSTOM MADE CHEMICALS LAB
New Product
COMMANDE
Développer tout / Réduire tout
,,2 components: 1000mg/l each of Si Ti in HNO3 5% HF 0.2% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15°C to 25°C,,,, 1 * 100 mL CMPLRM109672L1 1 * 100 mL
New Product
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou