15 components: 100mg/l each of As Ba Be Bi Cd Co Cr Cs Cu Ni Pb Rb Sr V Zn in HNO3 5% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15 1 * 100 mL
Fournisseur: CUSTOM MADE CHEMICALS LAB
15 components: 100mg/l each of As Ba Be Bi Cd Co Cr Cs Cu Ni Pb Rb Sr V Zn in HNO3 5% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15 1 * 100 mL
New Product
COMMANDE
Développer tout / Réduire tout
15 components: 100mg/l each of As Ba Be Bi Cd Co Cr Cs Cu Ni Pb Rb Sr V Zn in HNO3 5% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15° 1 * 100 mL CMPLRM109677L1 1 * 100 mL
New Product
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou