,,19 components: 100mg/l each of Ce Dy Er Eu Ga Gd Ho In La Lu Nd Pr Sm Tb Th Tm U Y Yb in HNO3 2% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store under normal laboratory conditions, at tem 1 * 100 mL
Fournisseur: CUSTOM MADE CHEMICALS LAB
New Product
COMMANDE
Développer tout / Réduire tout
,,19 components: 100mg/l each of Ce Dy Er Eu Ga Gd Ho In La Lu Nd Pr Sm Tb Th Tm U Y Yb in HNO3 2% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store under normal laboratory conditions, at tem 1 * 100 mL CMPLRM109681L1 1 * 100 mL
New Product
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou