,,14 components: 100ug/ml each of Al As Ca Cd Cr Cu Fe Hg K Mg Na Ni Pb Zn in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL
Fournisseur: CUSTOM MADE CHEMICALS LAB
,,14 components: 100ug/ml each of Al As Ca Cd Cr Cu Fe Hg K Mg Na Ni Pb Zn in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL
New Product
COMMANDE
Développer tout / Réduire tout
,,14 components: 100ug/ml each of Al As Ca Cd Cr Cu Fe Hg K Mg Na Ni Pb Zn in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL CMPLRM110592L1 1 * 100 mL
New Product
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou