Synonyms:
Saline, NaCl, Sea salt, Halite
VS1508.80G
CMPLVS1508.80GEA
0
EUR
InStock
CMPLVS1508.80G
Sodium chlorure, Solide
Sodium chlorure
Sodium chloride - certified reference material for standardisation of volumetric solutions.
Certifications: ISO 17034, ISO 17025, ISO 9001.
Informations de commande: 60 months shelf life - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.