Digi-Sense® Pro 4-Input Data Logging Thermocouple Thermometer
Fournisseur: CONTROL COMPANY
20250-20
COCO20250-20EA 0 EUR InStock
COCO20250-20
Digi-Sense® Pro 4-Input Data Logging Thermocouple Thermometer
Enregistreurs de données
Rugged, heavy-duty construction protects against knocks and jars.
  • Reliable measurements, guaranteed!
  • Precalibrated to NIST-traceable standards
  • Relative time stamp provides a time reference for major events
  • Electronic off set function allows compensation of probe errors
  • Infrared input port allows safe, noncontact measurements with optional IR wand
These professional type K thermometers provide accurate and reliable temperature measurement in an ergonomic design. Double-molded heavy-duty housing provides superior protection, making them ideal for tough applications. Plus the NIST-traceable precalibration means you get reliable measurements right out of the box, saving you time and money!
All models feature Min/Max/Avg, Hold, °F/°C or Kelvin (K) selectable units, relative time stamp on Min/Max/Avg, electronic offset function, temperature differential, backlit display, and auto power-off. Accept a wide variety of type K thermocouple probes with miniconnector.
Informations de commande: These meters also feature an infrared input that accepts an optional IR wand for noncontact measurements of objects that are difficult or dangerous to reach; order wand 20250-24 separately. The IR temperature reading can be used as a unique input when calculating temperature differential. The IR wand has a distance-to sight ratio of 8:1 and temperature range of –35 to 550 °C. For more information; contact your Avantor representative.
Information de livraison: Four Type K bead wire temperature probes, hard carrying case, USB cable, software, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité