Cleanroom Female Luer to Hose Barb Fittings, Straight Adapter
Fournisseur: Cole-Parmer
50109-85
COLE50109-85EA 0 EUR InStock
COLE50109-85
Cleanroom Female Luer to Hose Barb Fittings, Straight Adapter
Raccords et connecteurs
Cleanroom female Luer to hose barb fittings, straight adapter, ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of system.
  • Animal derivative free polypropylene
  • PVC-free ideal for use in bioprocess, life science, and pharmaceutical industries
  • Forms a leak-free luer connection with a quick simple turn
  • Single barb design tightly 'grips' the tubing
Manufactured in an ISO Class 7 certified cleanroom to minimise contamination by pollutants.
Certifications: Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité