Digital thermometer, bluetooth pocketherm with fold-out probe
Fournisseur: COMARK LIMITED
BT125
COMABT125EA 392 EUR InStock
COMABT125
Digital thermometer, bluetooth pocketherm with fold-out probe
Thermomètres Thermomètres numériques
The BT125 is designed for use in the kitchen and other food production areas. The Bluetooth Pocketherm thermometer transmits temperature data via Bluetooth to your Android or iOS phone or tablet.
  • Automates data collection and eliminates the need for handwritten temperature records
  • Probe can be angled for ergonomic use
  • Thermistor sensor
  • Auto switch off after 10 minutes to preserve battery life
  • Battery life of up to 1000 hours
  • Response time of 5 seconds
  • BioCote® antimicrobial
  • Waterproof to IP 65
  • Bluetooth ver 4.0 Low Energy (BLE) ranges up to 10 m
The stainless steel probe with a fast-responding 1,5 mm tip folds out to allow to choose the best angle to take different temperatures at different points in the kitchen such as prep, cooking or cooling.
Use with the Comark Kitchen Checks App to manage all the HACCP temperature recording. BioCote® has been officially recognised for its benefits within the food industry with HACCP International Certification.
Comark Kitchen Checks Dashboard allows the user to review that critical control checks have been undertaken and to assess the implications of any incidents outside of pre-set parameters.
Certifications: Contains FCC ID: P14BLGOO; UKAS (NIST equivalent); ISO 9001.
Information de livraison: One year warranty.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité