Pocket digital thermometer for dishwashers
Fournisseur: COMARK LIMITED
KM14
COMAKM14EA 531 EUR InStock
COMAKM14
Pocket digital thermometer for dishwashers
Thermomètres Thermomètres numériques
The KM14 dishwasher pocket thermometer incorporates a stainless steel penetration probe and can be placed inside a commercial dishwasher. A quick reading will show whether the dishwasher is operated at the correct temperature to sanitise the equipment.
  • Waterproof and field calibratable
  • Auto power off conserves battery life
  • BioCote® antimicrobial protection reduces the risk of cross-contamination
  • Combines accuracy with speed and durability
It is multi-purpose and can also be used as a conventional food penetration probe thermometer and is a top performing digital thermometer that combines accuracy, speed of response and durability. Also incorporates extended temperature range and max/min hold feature. It is designed to used for commercial dishwashers, so it is completely waterproof and has a ‘Max Hold’ feature that allows to verify the highest temperature record during the dishwasher cycle.
Certifications: NSF Certified.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité