Lingettes pour salles blanches, Polywipe-C
Fournisseur: CONTEC
PC-99 PC-1212
CONTPC-99EA 569 EUR InStock
CONTPC-99 CONTPC-1212
Lingettes pour salles blanches, Polywipe-C
Lingettes Lingettes pour salles propres Lingettes pour salles propres, sèches
Polywipe-C Wipes are made from a standardweight 100% knitted polyester that is knife cut to create a soft edge.
  • Soft edge is ideal for use on surfaces which are vulnerable to scratching
  • Low in both particles and extractables to reduce possibility of contamination
  • Good sorbency makes it compatible with many solvents
  • Abrasion and chemical resistant
  • Laundered and packed in an ISO Class 4
The soft edge is ideal for use on surfaces which are vulnerable to scratching. The wipe is chemical resistant and exceptionally low in particles and extractable residue making it ideal for critical cleaning.
Recommended for ISO Class 5 - 6 environments.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité