Cleaning swabs, double-ended, CONSTIX® SC-7
Fournisseur: CONTEC
SC-7
CONTSC-7EA 516 EUR InStock
CONTSC-7
Cleaning swabs, double-ended, CONSTIX® SC-7
Écouvillons Écouvillons de nettoyage
SC-7 sharp point, double-ended cotton swabs consist of high quality, long staple length cotton, tightly wrapped and shaped on both ends of high strength, wound paper handles.
  • Small, sharp pointed ends
  • High quality cotton fibres provide excellent particle entrapment and retention
  • Tightly wrapped head helps maintain low levels of extractable residue
  • Heads are held to the handles using a methyl-cellulose binder
The high quality cotton fibres provide exceptional sorbency of most solvents and solutions. The heads are very tightly wrapped and shaped using a methyl-cellulose binder which minimises fibre shedding. SC series cotton swabs are absorbent to most common solvents including isopropyl alcohol and acetone and are ideal for use with aqueous-based solutions.
These swabs are Sanyo Huby brand swabs imported by Contec.
Conditionnement: Swabs are packed 5000 per case.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité