Cleaning swabs, sealed foam, CONSTIX® SF-5
Fournisseur: CONTEC
SF-5
CONTSF-5EA 132 EUR InStock
CONTSF-5
Cleaning swabs, sealed foam, CONSTIX® SF-5
Écouvillons Écouvillons de nettoyage
This sealed foam swab consists of a polyurethane foam bonded to a flex-tip PP handle.
  • Flexible swab head tip provides good surface contact and controlled pressure to surfaces
  • Polyurethane foam head provides good particle entrapment and removal
  • No adhesives or binders used
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery or removal. The SF-5 swab is excellent for applying resins and adhesives.
Though compatible with many common solvents, the SF-5 is not recommended for use with ketones such as acetone or MEK.
Conditionnement: Swabs are packed 500 per case.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité