Cleaning swabs, sealed, CONSTIX® SF-9
Fournisseur: CONTEC
SF-9
CONTSF-9EA 89.9 EUR InStock
CONTSF-9
Cleaning swabs, sealed, CONSTIX® SF-9
Écouvillons Écouvillons de nettoyage
This sealed foam swab consists of polyurethane foam bonded to a polypropylene handle without the use of adhesives or binders.
  • Mini foam head enables good access to small features and difficult to reach surfaces
  • Polyurethane foam head provides good particle entrapment and removal
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery or removal. Excellent for applying resins and adhesives.
Though compatible with many common solvents, the SF-9 is not recommended for useMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité