Cleaning swabs, sealed, CONSTIX® SP-11
Fournisseur: CONTEC
SP-11
CONTSP-11EA 108 EUR InStock
CONTSP-11
Cleaning swabs, sealed, CONSTIX® SP-11
Écouvillons Écouvillons de nettoyage
The SP-11 sealed polyester swab consists of 100% knitted polyester fabric that’s thermally bonded to a rigid PP handle.
  • Durable and absorbent
  • Features extremely low particles and residue levels
  • Polyester head provides enhanced sorbency, particle entrapment and removal
  • Polyester head makes the swab compatible with most common solvents, such as alcohols and ketones
  • Flexible tip enables access to very small areas and components
  • PP handle has a thin tip
The swab head is made of cleanroom laundered; double-layer knitted microfibre polyester material. The polyester head is thermally sealed and attached, also constructed without adhesives or binders.
Conditionnement: Swabs are packed 500 per case.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité