Cleaning swabs, sealed, CONSTIX® SP-8
Fournisseur: CONTEC
SP-8
CONTSP-8EA 70.9 EUR InStock
CONTSP-8
Cleaning swabs, sealed, CONSTIX® SP-8
Écouvillons Écouvillons de nettoyage
The SP-8 sealed polyester swab consists of 100% knitted polyester fabric that is thermally bonded to a PP handle with a large paddle.
  • Durable and absorbent
  • Features extremely low particles and residue levels
  • Polyester head provides enhanced sorbency, particle entrapment and removal
  • Large paddle swab head provides broader surface coverage in critical applications
The swab head is made of cleanroom laundered; double-layer knitted microfibre polyester material. The polyester head is constructed without adhesives or binders, which makes the swab compatible with most common solvents, such as alcohols and ketones.
Conditionnement: Swabs are packed 100 per case.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité