Cleanroom wipes, pre-saturated, sterile, SATWipes HydroPure
Fournisseur: CONTEC
SSWC16HP
CONTSSWC16HPEA 825 EUR InStock
CONTSSWC16HP
Cleanroom wipes, pre-saturated, sterile, SATWipes HydroPure
Lingettes Lingettes pour salles propres Lingettes pour salles propres, présaturées
Contec HydroPure is a blend of 6% hydrogen peroxide and purified water. Contec HydroPure is BPR authorised at EU level and efficacious against bacteria, fungi, moulds, and yeasts.
  • Impregnated wipes at point-of-use provide convenient and controlled application of disinfectant
  • Hydrogen Peroxide breaks down to water & oxygen leaving no residue
  • 100 polyester / cellulose wipes in a canister
  • Filtered to 0.2 microns in a Grade B cleanroom
  • Ideal for a Grade A / B areas
  • Not classed as corrosive
  • Double bagged in linear tear packaging
Contec HydroPure leaves little-to-no residue making it ideal for use in critical areas. Contec HydroPure is provided as a point-of-use SATWIPES system. Each double bagged pack contains a canister of 100 x 54gsm Amplitude Theta polyester/ cellulose dry wipes and a 500ml capped bottle of Contec HydroPure.
Simply add 500 ml of HydroPure to the dry roll, 5 mins before use, and use the impregnated wipes within 7 days.
Contec HydroPure is 0.2 micron filtered, and filled in a Grade A environment. The sterile product is filtered into pre-irradiated components. The dry wipe canister is irradiated at no less than 25 kGy
Attention: Utilisez les biocides en toute sécurité. Toujours lire l'étiquette et les informations du produit avant utilisation.
*Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité