Cleanroom wipes, pre-saturated, SATWIPES® Theta
Fournisseur: CONTEC
SWC10004
CONTSWC10004EA 280 EUR InStock
CONTSWC10004
Cleanroom wipes, pre-saturated, SATWIPES® Theta
Lingettes Lingettes pour salles propres Lingettes pour salles propres, présaturées
Presaturated wipes can offer many benefits over bottled alcohol and dry wipes for cleanroom use. Contec’s sterile SATWIPES Theta with 70% denatured ethanol consist of a perforated roll of presaturated wipes which are dispensed through a convenient centre pull canister.
  • Made of 55% cellulose and 45% polyester creating a highly sorbent and durable wipe
  • Presaturated wipes eliminate the need for trigger spray bottles
  • Low levels of particles and fibres
  • Resealable canister prevents wipes drying out during use
  • Linear tear bags are easy to open even when wearing gloves
These sterile wipes are ideal when larger numbers of wipes are needed such as during transfer disinfection.
Conditionnement: Double bagged for ease of use in pharmaceutical cleanrooms.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité