Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

L'expérience en ligne d'Avantor évolue pour mieux vous accompagner ! À compter du 4 avril 2025, nos clients seront migrés vers une nouvelle plateforme pour une expérience d'achat en ligne simplifiée.

NAO™ Baskets for Rapid and Efficient DNA Recovery

Fournisseur: Copan

3002C 3102CS01 4103CS01 30U013D01
COPA3002CEA 872 EUR InStock
COPA3002C COPA3102CS01 COPA4103CS01 COPA30U013D01
NAO™ Baskets for Rapid and Efficient DNA Recovery
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents

NAO® Basket for rapid and efficient DNA recovery. This is a semi-permeable system designed for releasing and concentrating human DNA from swab samples or other specimens during the extraction step. Lysis in the NAO® Basket: NAO® Basket retains the lysis buffer during the lysis step of the forensic sample.



  • Application: DNA rapid recovory

  • Lyse and centrifuge in the basket: innovative product design allows the passage of DNA from samples into the tube using a tight leak-proof design that can endure vortexing, centrifugations, and various storage conditions

  • Multiple configurations: NAO® Basket are available in several formats as single pouches or bulk packages, both with 1,5 ml or 2 ml vials
  • Compatibility: NAO® Basket is compatible with commercial Nucleic Extraction Platforms
  • Forensic DNA grade: ISO 18385:2016 compliance minimises the risk of detectable human nuclear DNA contamination in products used by the global forensic community


Upon centrifugation on a high-speed centrifuge, valves in the bottom of the NAO® Basket open up and allow the sample lysate to pass through. The lysate containing the nucleic acids is efficiently recovered and collected in the provided collection tube, whereas the substrate remains in the NAO® Basket.



How to use NAO® Basket:
1. Break the FLOQSwabs® at the breaking point into the NAO® Basket.
2. Add the lysis buffer and incubate as per extraction kit instructions.
3. Upon centrifugation, the sample lysate passes through the basket and is available for testing at the bottom of the tube. Improve DNA yield from challenging samples:
The combination of NAO® Basket and FLOQSwabs® provides more than 90% DNA recovery of the reference sample collected by the swab, particularly critical for crime scene evidence testing. The 20 mm breaking point designed on the 4N6FLOQSwabs® is compatible with the NAO® Basket.



Certifications: Certification: ISO 18385:2016. The validated EO TREATMENT assesses the degradation of possible human DNA residuals. Free of amplifiable human DNA, detectable and amplifiable DNase, and RNase. Personnel STR database.

Attention: The NAO® Basket is not intended for diagnostic purposes.


Commander maintenant

 

COMMANDE


Appel de prix
Le stock de cet article est limité mais peut être disponible dans un entrepôt proche de vous. Merci de vous assurer que vous êtes connecté sur le site afin que le stock disponible soit affiché. Si l'call est toujours affiché et vous avez besoin d'aide, s'il vous plaît appelez-nous au 016 385 011
Le stock de cet article est limité mais peut être disponible dans un entrepôt proche de vous. Merci de vous assurer que vous êtes connecté sur le site afin que le stock disponible soit affiché. Si l'call est toujours affiché et vous avez besoin d'aide, s'il vous plaît appelez-nous au 016 385 011
Ces articles ne peuvent être ajoutés au Panier. Veuillez contacter votre service client ou envoyer un e-mail à vwr.be@vwr.com
Une documentation supplémentaire peut être nécessaire pour l'achat de cet article. Un représentant de VWR vous contactera si nécessaire.
Ce produit a été bloqué par votre organisation. Contacter votre service d'achat pour plus d'informations.
Le produit original n'est plus disponible. Le remplacement représenté est disponible
Les produits marqués de ce symbole ne seront bientôt plus disponibles - vente jusqu'à épuisement de stock. Des alternatives peuvent être disponibles en recherchant le code article VWR indiqué ci-dessus. Si vous avez besoin d'une assistance supplémentaire, veuillez contacter notre Service Clientèle au 016 385 011.
Collapse All
Désignation Cdt Numéro de catalogue VWR Quantité
NAO™ basket in bulk bag, 250×6, Packet/Batch 1.500 COPA3002C
NAO™ Basket with 2 ml brand/treff cuvette, in peel pouch, 100×6, Individually wrapped 600 COPA3102CS01
NAO™ basket with 2 ml brand/tref cuvette with 4n6FLOQSwabs® genetics regular size tip, 20 mm breaking point, 100×6, Individually wrapped - COPA4103CS01
NAO™ basket with 1,5 ml cuvette in peel pouch, 100×6, Individually wrapped 600 COPA30U013D01

Générer des étiquettes code à barres. Format des étiquettes code à barres.: 
Créer une page catalogue sous forme pdf

claquement

Veuillez sélectionner l'article

Entrer les détails

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité

Ce site utilise des cookies, en provenance de VWR ou de ses partenaires, afin de collecter des informations statistiques sur votre navigation et vous proposer des contenus en accord avec vos préférences, générés en fonction de vos habitudes de navigation. En poursuivant la consultation de ce site, vous approuvez l’utilisation de ces cookies.

Pour en savoir plus, consultez notre Politique de Confidentialité..