Beakers, low form, borosilicate glass, PYREX®
Fournisseur: Corning
1000-30
CORN1000-30EA 0 EUR InStock
CORN1000-30
Beakers, low form, borosilicate glass, PYREX®
Béchers
Made of PYREX® borosilicate glass.
  • PYREX®, Soda-lime glass, with spout
  • Very good chemical resistance
  • Good temperature resistance
Manufactured with uniform wall thickness to offer optimum balance between thermal shock resistance and mechanical strength.
Sizes 250 ml and larger have a double graduated metric scale to indicate approximate content. The 10 ml size is not graduated. All sizes have an extra large marking spot.
Certifications: Meet ASTM E960 specifications.
Informations de commande: The variety pack includes one of each of the following beakers: 50, 100, 250, 600 and 1000 ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité